Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107951.1 | NP_001101421.1 | 1 - 2 | 287 | 113 | |
XM_006238589.1 | XP_006238651.1 | 1 - 2 | 287 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |