Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106682.1 | NP_001100152.1 | 1 - 2 | 44 | 82 | |
GenBank mRNA | BC100634.1 | - | 1 - 2 | 44 | 82 | |
BC168679.1 | - | 1 - 2 | 44 | 5621187 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |