Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750019.3 | XP_003750067.1 | 2 - 3 | 166 | 73 | |
XM_006238771.2 | XP_006238833.1 | 2 - 3 | 166 | 73 | ||
XM_006238772.2 | XP_006238834.1 | 2 - 3 | 166 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |