Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085369.1 | NP_001078838.1 | 17 - 18 | 1828 | 106 | |
NM_134472.3 | NP_608302.2 | 16 - 17 | 1713 | 106 | ||
XM_006238632.2 | XP_006238694.1 | 17 - 18 | 1880 | 106 | ||
XM_006238633.1 | XP_006238695.1 | 16 - 17 | 1644 | 106 | ||
XM_006238634.2 | XP_006238696.1 | 16 - 17 | 2176 | 106 | ||
XM_006238635.2 | XP_006238697.1 | 17 - 18 | 2053 | 106 | ||
XM_006238636.2 | XP_006238698.1 | 17 - 18 | 2049 | 106 | ||
GenBank mRNA | CO403678.1 | - | 4 - 5 | 482 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |