Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhcr24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080148.1 | NP_001073617.1 | 6 - 7 | 1024 | 102 | |
GenBank mRNA | AY310319.1 | - | 6 - 7 | 1024 | 102 | |
AY921220.2 | - | 6 - 7 | 1086 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |