Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025411.1 | NP_001020582.1 | 4 - 5 | 974 | 119 | |
GenBank mRNA | BC082010.1 | - | 4 - 5 | 972 | 7131770 | 119 |
BC097438.1 | - | 4 - 5 | 974 | 7455678 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |