Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025411.1 | NP_001020582.1 | 6 - 7 | 1426 | 74 | |
GenBank mRNA | BC082010.1 | - | 6 - 7 | 1424 | 7131770 | 74 |
BC097438.1 | - | 6 - 7 | 1426 | 7455678 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |