Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024351.2 | NP_077327.1 | 6 - 7 | 1399 | 67 | |
GenBank mRNA | BC061547.1 | - | 6 - 7 | 1406 | 6921606 | 67 |
BC098914.1 | - | 6 - 7 | 1351 | 7104230 | 67 | |
FQ213866.1 | - | 2 - 3 | 415 | 67 | ||
FQ226406.1 | - | 2 - 3 | 334 | 67 | ||
FQ231618.1 | - | 1 - 2 | 46 | 67 | ||
FQ232261.1 | - | 2 - 3 | 334 | 67 | ||
M11942.1 | - | 6 - 7 | 1370 | 67 |
Gene Symbol: LOC689955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764620.1 | XP_008762842.1 | 1 | 1401 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |