Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080756.1 | NP_001074225.1 | 6 - 7 | 590 | 94 | |
XM_006243310.2 | XP_006243372.1 | 6 - 7 | 744 | 94 | ||
XM_006243311.2 | XP_006243373.1 | 6 - 7 | 744 | 94 | ||
GenBank mRNA | AY952140.1 | - | 6 - 7 | 554 | 94 | |
EF187255.1 | - | 6 - 7 | 590 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |