Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cln6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191794.1 | NP_001178723.1 | 2 - 3 | 209 | 60 | |
XM_006243238.2 | XP_006243300.1 | 3 - 4 | 299 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |