Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017281.1 | NP_058977.1 | 2 - 3 | 98 | 85 | |
GenBank mRNA | BC063170.1 | - | 2 - 3 | 118 | 6921964 | 85 |
X53304.1 | - | 2 - 3 | 98 | 85 | ||
X55986.1 | - | 2 - 3 | 54 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |