Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009697.1 | NP_001009697.1 | 5 - 6 | 724 | 141 | |
GenBank mRNA | BC088470.1 | - | 5 - 6 | 724 | 7128152 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |