Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004247.1 | NP_001004247.1 | 4 - 5 | 484 | 82 | |
XM_006243155.2 | XP_006243217.1 | 4 - 5 | 452 | 82 | ||
XM_006243156.2 | XP_006243218.1 | 4 - 5 | 504 | 82 | ||
GenBank mRNA | BC079221.1 | - | 4 - 5 | 484 | 7131980 | 82 |
FQ212102.1 | - | 4 - 5 | 709 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |