Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257079.1 | XP_006257141.1 | 4 - 5 | 616 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus Probe spans exons |