Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173151.1 | NP_775174.1 | 2 - 3 | 229 | 91 | |
XM_006257016.2 | XP_006257078.1 | 2 - 3 | 190 | 91 | ||
XM_006257017.2 | XP_006257079.1 | 2 - 3 | 597 | 91 | ||
XM_008773254.1 | XP_008771476.1 | 2 - 3 | 310 | 91 | ||
GenBank mRNA | AF190256.1 | - | 2 - 3 | 229 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |