Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212510.1 | NP_997675.1 | 2 - 3 | 232 | 106 | |
GenBank mRNA | BC058498.1 | - | 2 - 3 | 232 | 6919102 | 106 |
FQ209874.1 | - | 1 - 2 | 73 | 106 | ||
FQ209881.1 | - | 2 - 3 | 226 | 106 | ||
FQ209956.1 | - | 2 - 3 | 232 | 106 | ||
FQ216877.1 | - | 2 - 3 | 225 | 106 | ||
FQ221165.1 | - | 2 - 3 | 227 | 106 | ||
FQ221243.1 | - | 2 - 3 | 226 | 106 | ||
FQ221413.1 | - | 2 - 3 | 226 | 106 | ||
FQ221458.1 | - | 2 - 3 | 226 | 106 | ||
FQ221831.1 | - | 2 - 3 | 106 | |||
FQ222162.1 | - | 2 - 3 | 226 | 106 | ||
FQ222289.1 | - | 2 - 3 | 226 | 106 | ||
FQ222457.1 | - | 2 - 3 | 226 | 106 | ||
FQ222474.1 | - | 2 - 3 | 230 | 106 | ||
FQ222662.1 | - | 2 - 3 | 226 | 106 | ||
FQ223818.1 | - | 2 - 3 | 227 | 106 | ||
FQ228440.1 | - | 2 - 3 | 226 | 106 | ||
X14181.1 | - | 2 - 3 | 226 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |