Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053580.2 | NP_446032.2 | 9 - 10 | 1510 | 136 | |
XM_006252953.2 | XP_006253015.1 | 9 - 10 | 1548 | 136 | ||
XM_006252954.1 | XP_006253016.1 | 9 - 10 | 1496 | 136 | ||
XM_008771156.1 | XP_008769378.1 | 9 - 10 | 1435 | 136 | ||
XM_008771157.1 | XP_008769379.1 | 9 - 10 | 1594 | 136 | ||
GenBank mRNA | BC074014.1 | - | 9 - 10 | 1510 | 7104661 | 136 |
U89529.1 | - | 9 - 10 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |