Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271055.1 | NP_001257984.1 | 6 - 7 | 819 | 71 | |
XM_006252897.2 | XP_006252959.1 | 7 - 8 | 889 | 71 | ||
GenBank mRNA | BC089056.1 | - | 6 - 7 | 746 | 71 | |
FQ214027.1 | - | 6 - 7 | 773 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |