Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mef2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017507.1 | NP_001017507.1 | 3 - 4 | 321 | 75 | |
XM_006252927.2 | XP_006252989.1 | 3 - 4 | 641 | 75 | ||
GenBank mRNA | BC079361.1 | - | 3 - 4 | 321 | 7109152 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |