Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mef2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017507.1 | NP_001017507.1 | 4 - 5 | 460 | 86 | |
XM_006252927.2 | XP_006252989.1 | 4 - 5 | 780 | 86 | ||
GenBank mRNA | BC079361.1 | - | 4 - 5 | 460 | 7109152 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |