Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106962.1 | NP_001100432.1 | 5 - 6 | 592 | 146 | |
XM_006256858.2 | XP_006256920.1 | 5 - 6 | 946 | 146 | ||
XM_008773166.1 | XP_008771388.1 | 5 - 6 | 997 | 146 | ||
GenBank mRNA | FQ216453.1 | - | 5 - 6 | 698 | 146 | |
FQ224911.1 | - | 5 - 6 | 651 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |