Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107865.1 | NP_001101335.1 | 6 - 7 | 762 | 92 | |
XM_006236673.2 | XP_006236735.1 | 7 - 8 | 1071 | 92 | ||
XM_006236674.2 | XP_006236736.1 | 7 - 8 | 1068 | 92 | ||
XM_006236675.2 | XP_006236737.1 | 6 - 7 | 1029 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |