Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setmar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025048.1 | NP_001020219.1 | 1 - 2 | 174 | 98 | |
GenBank mRNA | BC088181.1 | - | 1 - 2 | 174 | 7364937 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |