Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012641.1 | NP_036773.1 | 4 - 5 | 400 | 96 | |
GenBank mRNA | FQ232485.1 | - | 4 - 5 | 392 | 96 | |
M18962.1 | - | 3 - 4 | 376 | 96 | ||
M62930.1 | - | 4 - 5 | 400 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons |