Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243172.2 | XP_006243234.1 | 6 - 7 | 1262 | 57 | |
XM_006243174.2 | XP_006243236.1 | 5 - 6 | 923 | 57 | ||
GenBank mRNA | FQ226464.1 | - | 1 - 2 | 122 | 57 | |
X99337.1 | - | 5 - 6 | 952 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |