Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp11a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017286.2 | NP_058982.1 | 2 - 3 | 424 | 115 | |
GenBank mRNA | BC089100.1 | - | 2 - 3 | 449 | 7307104 | 115 |
DQ515795.1 | - | 2 - 3 | 411 | 115 | ||
J05156.1 | - | 2 - 3 | 444 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |