Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106813.1 | NP_001100283.1 | 1 - 2 | 49 | 88 | |
XM_008766122.1 | XP_008764344.1 | 1 - 2 | 154 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |