Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_236272.6 | XP_236272.4 | 4 - 5 | 704 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus Probe spans exons |