Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014093.1 | NP_001014115.1 | 3 - 4 | 639 | 83 | |
XM_006243257.2 | XP_006243319.1 | 4 - 5 | 1027 | 83 | ||
XM_006243258.2 | XP_006243320.1 | 4 - 5 | 795 | 83 | ||
GenBank mRNA | AY387095.1 | - | 891 | 83 | ||
BC085908.1 | - | 3 - 4 | 639 | 7096186 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |