Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022005.2 | NP_071288.1 | 3 - 4 | 319 | 102 | |
XM_008766171.1 | XP_008764393.1 | 3 - 4 | 348 | 99 | ||
GenBank mRNA | AB030908.1 | - | 3 - 4 | 133 | 102 | |
AF142439.1 | - | 3 - 4 | 129 | 99 | ||
BC072528.1 | - | 3 - 4 | 125 | 7096832 | 102 | |
CB741544.1 | - | 3 - 4 | 341 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |