Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044253.1 | NP_001037718.1 | 1 - 2 | 541 | 77 | |
XM_006243271.2 | XP_006243333.1 | 1 - 2 | 1478 | 77 | ||
XM_008766324.1 | XP_008764546.1 | 2 - 3 | 1197 | 77 | ||
XM_008766325.1 | XP_008764547.1 | 2 - 3 | 1169 | 77 | ||
XM_008766326.1 | XP_008764548.1 | 2 - 3 | 1128 | 77 | ||
XM_008766327.1 | XP_008764549.1 | 2 - 3 | 1100 | 77 | ||
GenBank mRNA | DQ489562.1 | - | 1 - 2 | 541 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |