Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108143.2 | NP_001101613.2 | 1 - 2 | 243 | 130 | |
GenBank mRNA | BC166987.1 | - | 1 - 2 | 243 | 7442233 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |