Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108143.2 | NP_001101613.2 | 2 - 3 | 411 | 103 | |
GenBank mRNA | BC166987.1 | - | 2 - 3 | 411 | 7442233 | 103 |
FQ229819.1 | - | 318 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |