Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cilp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108161.1 | NP_001101631.1 | 7 - 8 | 1172 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |