Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173426.1 | NP_001166897.1 | 6 - 7 | 560 | 107 | |
XM_006243274.2 | XP_006243336.1 | 6 - 7 | 2246 | 107 | ||
XM_008766329.1 | XP_008764551.1 | 5 - 6 | 669 | 107 | ||
GenBank mRNA | BC169038.1 | - | 6 - 7 | 560 | 9090232 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |