Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017085.2 | NP_058781.2 | 3 - 4 | 903 | 82 | |
GenBank mRNA | EF091858.1 | - | 2 - 3 | 190 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Inventoried Probe spans exons |