Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053660.2 | NP_446112.1 | 1 - 2 | 126 | 136 | |
GenBank mRNA | BC105759.1 | - | 1 - 2 | 119 | 5623275 | 136 |
CA340326.1 | - | 1 - 2 | 123 | 136 | ||
CB616034.1 | - | 1 - 2 | 126 | 136 | ||
FQ214425.1 | - | 1 - 2 | 110 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |