Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpjl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108604.1 | NP_001102074.1 | 8 - 9 | 1009 | 60 | |
XM_006235598.2 | XP_006235660.1 | 7 - 8 | 951 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |