Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130399.2 | NP_569083.1 | 9 - 10 | 917 | 76 | |
GenBank mRNA | AB059655.1 | - | 9 - 10 | 841 | 76 | |
BC088116.1 | - | 9 - 10 | 917 | 7384286 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |