Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168543.1 | NP_001162015.1 | 3 - 4 | 451 | 64 | |
XM_006235535.2 | XP_006235597.1 | 4 - 5 | 509 | 64 | ||
XM_008762476.1 | XP_008760698.1 | 4 - 5 | 503 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |