Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168543.1 | NP_001162015.1 | 8 - 9 | 960 | 101 | |
XM_006235535.2 | XP_006235597.1 | 9 - 10 | 1018 | 101 | ||
XM_008762476.1 | XP_008760698.1 | 9 - 10 | 1012 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |