Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106542.1 | NP_001100012.1 | 2 - 3 | 176 | 98 | |
GenBank mRNA | FQ222472.1 | - | 2 - 3 | 167 | 98 | |
FQ234128.1 | - | 2 - 3 | 170 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |