Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106544.1 | NP_001100014.1 | 2 - 3 | 284 | 93 | |
GenBank mRNA | FQ214139.1 | - | 2 - 3 | 267 | 93 | |
FQ216824.1 | - | 2 - 3 | 267 | 93 | ||
FQ219919.1 | - | 2 - 3 | 267 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |