Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnttip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134400.2 | NP_599227.2 | 6 - 7 | 549 | 92 | |
XM_006235512.2 | XP_006235574.1 | 6 - 7 | 568 | 92 | ||
XM_006235513.2 | XP_006235575.1 | 6 - 7 | 517 | 92 | ||
GenBank mRNA | AF348701.1 | - | 6 - 7 | 517 | 92 | |
BC100640.1 | - | 6 - 7 | 549 | 7929876 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |