Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011959.1 | NP_001011959.1 | 10 - 11 | 1237 | 125 | |
XM_006235534.1 | XP_006235596.1 | 11 - 12 | 1495 | 125 | ||
GenBank mRNA | BC078934.1 | - | 10 - 11 | 1237 | 7108104 | 125 |
FQ219992.1 | - | 3 - 4 | 290 | 125 | ||
FQ229081.1 | - | 10 - 11 | 1391 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |