Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025768.1 | NP_001020939.1 | 10 - 11 | 1034 | 102 | |
XM_006235375.2 | XP_006235437.1 | 10 - 11 | 1344 | 102 | ||
XM_006235376.2 | XP_006235438.1 | 10 - 11 | 2015 | 102 | ||
XM_006235377.2 | XP_006235439.1 | 10 - 11 | 2107 | 102 | ||
GenBank mRNA | BC098027.1 | - | 10 - 11 | 1034 | 7456020 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |