Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025768.1 | NP_001020939.1 | 9 - 10 | 943 | 98 | |
XM_006235375.2 | XP_006235437.1 | 9 - 10 | 1253 | 98 | ||
XM_006235376.2 | XP_006235438.1 | 9 - 10 | 1924 | 98 | ||
XM_006235377.2 | XP_006235439.1 | 9 - 10 | 2016 | 98 | ||
GenBank mRNA | BC098027.1 | - | 9 - 10 | 943 | 7456020 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |