Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024381.2 | NP_077357.2 | 12 - 13 | 1051 | 68 | |
XM_006256987.2 | XP_006257049.1 | 13 - 14 | 1103 | 68 | ||
XM_006256988.2 | XP_006257050.1 | 12 - 13 | 1085 | 68 | ||
XM_008773274.1 | XP_008771496.1 | 14 - 15 | 1168 | 68 | ||
XM_008773275.1 | XP_008771497.1 | 13 - 14 | 1150 | 68 | ||
XM_008773276.1 | XP_008771498.1 | 14 - 15 | 1168 | 68 | ||
XM_008773278.1 | XP_008771500.1 | 14 - 15 | 1167 | 68 | ||
GenBank mRNA | CO564445.1 | - | 3 - 4 | 184 | 68 | |
D16102.1 | - | 12 - 13 | 1054 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |