Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013004.1 | NP_037136.1 | 8 - 9 | 1410 | 134 | |
GenBank mRNA | AJ001637.1 | - | 8 - 9 | 1410 | 134 |
Gene Symbol: LOC102554012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762111.1 | XP_008760333.1 | 2 - 3 | 1287 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Rattus norvegicus Probe spans exons |