Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033963.1 | NP_001029135.1 | 7 - 8 | 1204 | 92 | |
GenBank mRNA | BC091203.1 | - | 7 - 8 | 1204 | 7380393 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |